Learn More
Greiner Bio-One 96-well CrystalQuick™ Crystallisation Microplates
For automatised applications. Greiner Bio-One 96-Well CrystalQuick™ Crystallisation Microplates are optimised for sealing with VIEWseal adhesive film.
Marca: Greiner Bio-One 609801
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Non-sterile
- Polystyrene
- Good optical properties
- With crystallisation well per reservoir
- Alphanumeric well coding
- With CrystalDrop™ lid, experiments are possible with sitting drop and hanging drop metho
Specifica
Standard | |
Inner bags of 10 | |
LBR (low birefringence) | |
4.1 μL | |
Non-sterile | |
Sitting drop applications with different well profiles and material properties | |
40/Cs. |
8 x 12 | |
Clear | |
Polystyrene | |
96 | |
Square, Flat Bottom (F-well) | |
40 | |
14.4 mm |