Learn More
Cytiva (formerly Pall Lab) A/E Glass Fiber Disc Filters
Superior grade filters for a variety of biological and environmental methods.
Marca: Cytiva (formerly Pall Lab) 61633
Dettagli aggiuntivi : Peso : 0.15200kg
Descrizione
- Type A/E meets the requirements for suspended solids testing, as described in Standard Methods for the Examination of Water and Wastewater, current edition.
- Reduces filtration costs and premature clogging when filtering difficult-to-filter or highly contaminated solutions.
Specifica
60 L/min/cm2 | |
1 μm | |
100/Pk. | |
102 mm |
Borosilicate glass without binder | |
Non-sterile | |
550°C |