missing translation for 'onlineSavingsMsg'
Learn More

AASDHPPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11244-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898183

  • 325.38€ / 150µL

Spedizione stimata: 03-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The protein encoded by this gene is similar to Saccharomyces cerevisiae LYS5, which is required for the activation of the alpha-aminoadipate dehydrogenase in the biosynthetic pathway of lysine. Yeast alpha-aminoadipate dehydrogenase converts alpha-biosynthetic-aminoadipate semialdehyde to alpha-aminoadipate. It has been suggested that defects in the human gene result in pipecolic acidemia.
Specifica

Specifica

AASDHPPT
Polyclonal
Unconjugated
AASDHPPT
AASD PPT, AASDHPPT, CGI 80, HAH P, HSPC223, LYS2, LYS5, LYS5 ortholog, x0005
Rabbit
Antigen Affinity Chromatography
RUO
300328, 60496, 67618
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9CQF6, Q9NRN7
AASDHPPT
AASDHPPT Fusion Protein Ag1763
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali