missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18179-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ABCD2 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown; however this protein is speculated to function as a dimerization partner of ABCD1 and/or other peroxisomal ABC transporters.Specifica
| ABCD2 | |
| Polyclonal | |
| Unconjugated | |
| ABCD2 | |
| ABC39, ABCD2, Adrenoleukodystrophy like 1, ALD1, ALDL1, ALDR, ALDRP, hALDR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 225, 26874, 84356 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q61285, Q9QY44, Q9UBJ2 | |
| Abcd2 | |
| ABCD2 Fusion Protein Ag12872 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto