missing translation for 'onlineSavingsMsg'
Learn More

ABCD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifica

Antigene ABCD2
Concentrazione 0.3 mg/mL
Applicazioni Western Blot, Immunoprecipitation
Classificazione Polyclonal
Coniugato Unconjugated
Visualizza altre specifiche

Products 2
Product Code Brand Quantità Price Quantity & Availability  
Product Code Brand Quantità Price Quantity & Availability  
16818244
missing translation for 'viewDocuments'
Proteintech
18179-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16808244
missing translation for 'viewDocuments'
Proteintech
18179-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Description

Description

ABCD2 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown; however this protein is speculated to function as a dimerization partner of ABCD1 and/or other peroxisomal ABC transporters.
Specifications

Specifications

ABCD2
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Rat, Human
Q61285, Q9QY44, Q9UBJ2
225, 26874, 84356
ABCD2 Fusion Protein Ag12872
Primary
-20°C
ABCD2
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ABC39, ABCD2, Adrenoleukodystrophy like 1, ALD1, ALDL1, ALDR, ALDRP, hALDR
Abcd2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.