missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13578-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. The ABC transporter ABCG1 (ABC8) is expressed on the cell surface and in intracellular compartments of cholesterol-laden macrophages and is induced in monocyte-derived macrophages during cholesterol influx mediated by acetylated low-density lipoprotein. In contrast, lipid efflux in cholesterol-laden macrophages, mediated by the cholesterol acceptor HDL(3), suppresses the expression of ABCG1.Specifica
| ABCG1 | |
| Polyclonal | |
| Unconjugated | |
| ABCG1 | |
| ABC8, ABCG1, White protein homolog, WHITE1, WHT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11307, 85264, 9619 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P45844, Q64343 | |
| Abcg1 | |
| ABCG1 Fusion Protein Ag4479 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto