missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifica
| Antigene | ABCG1 |
|---|---|
| Concentrazione | 0.24 mg/mL |
| Applicazioni | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Produktkod | Brand | Quantità | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantità | Pris | Kvantitet och tillgänglighet | |||||
|
16841864
|
Proteintech
13578-1-AP-20UL |
20 μL |
145.00€
20µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
16831864
|
Proteintech
13578-1-AP-150UL |
150 μL |
410.00€
150µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Beskrivning
ATP-binding cassette (ABC) transporter genes are involved in the regulation of the amount of dietary cholesterol retained in the body. The ABC transporter ABCG1 (ABC8) is expressed on the cell surface and in intracellular compartments of cholesterol-laden macrophages and is induced in monocyte-derived macrophages during cholesterol influx mediated by acetylated low-density lipoprotein. In contrast, lipid efflux in cholesterol-laden macrophages, mediated by the cholesterol acceptor HDL(3), suppresses the expression of ABCG1.Specifikationer
| ABCG1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P45844, Q64343 | |
| 11307, 85264, 9619 | |
| ABCG1 Fusion Protein Ag4479 | |
| Primary | |
| -20°C | |
| ABCG1 |
| 0.24 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ABC8, ABCG1, White protein homolog, WHITE1, WHT1 | |
| Abcg1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel