Learn More
ABHD5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 12201-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16899763
  • 140.00€ / 20µL
Spedizione stimata: 16-04-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
The protein encoded by this gene belongs to a large family of proteins defined by an alpha/beta hydrolase fold, and contains three sequence motifs that correspond to a catalytic triad found in the esterase/lipase/thioesterase subfamily. It differs from other members of this subfamily in that its putative catalytic triad contains an asparagine instead of the serine residue. Mutations in this gene have been associated with Chanarin-Dorfman syndrome, a triglyceride storage disease with impaired long-chain fatty acid oxidation.
Specifica
Specifica
ABHD5
Polyclonal
Unconjugated
ABHD5
ABHD5, CDS, CGI58, IECN2, NCIE2
Rabbit
Antigen Affinity Chromatography
RUO
51099, 67469
-20°C
Liquid
Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WTS1, Q9DBL9
ABHD5
ABHD5 Fusion Protein Ag2840
20 μL
Primary
Mouse, Human