Learn More
ABHD6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567277
Dettagli aggiuntivi : Peso : 0.00001kg
Descrizione
Immunogen sequence: GKQDQVLDVSG ADMLAKSIAN CQVELLENCG HSVVMERPRK TAKLIIDFLA SVHNTDNNKK LD Highest antigen sequence identity to the following orthologs - mouse 86%, rat 87%.
ABHD6 has 2-arachidonoylglycerol hydrolase activity. ABHD6 may be a regulator of endocannabinoid signaling pathways.Specifica
ABHD6 | |
Polyclonal | |
Unconjugated | |
ABHD6 | |
0610041D24Rik; 2-arachidonoylglycerol hydrolase; AA673485; Abhd6; abhydrolase domain containing 6; abhydrolase domain containing 6, acylglycerol lipase; abhydrolase domain-containing protein 6; AV065425; lipase protein; monoacylglycerol lipase ABHD6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57406 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BV23 | |
ABHD6 | |
Recombinant Human ABHD6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only