missing translation for 'onlineSavingsMsg'
Learn More

ACKR2 Mouse anti-Human, Clone: 1A3A2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60046-1-IG-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896995

  • 121.38€ / 20µL

Spedizione stimata: 10-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes a beta chemokine receptor, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptor-mediated signal transduction are critical for the recruitment of effector immune cells to the inflammation site. This gene is expressed in a range of tissues and hemopoietic cells. The expression of this receptor in lymphatic endothelial cells and overexpression in vascular tumors suggested its function in chemokine-driven recirculation of leukocytes and possible chemokine effects on the development and growth of vascular tumors. This receptor appears to bind the majority of beta-chemokine family members; however, its specific function remains unknown. This gene is mapped to chromosome 3p21.3, a region that includes a cluster of chemokine receptor genes.
Specifica

Specifica

ACKR2
Monoclonal
0.83 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O00590
ACKR2
ACKR2 Fusion Protein Ag1941
20 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin)
1A3A2
Unconjugated
ACKR2
ACKR2, C C chemokine receptor D6, CCBP2, CCR10, CCR9, chemokine binding protein 2, Chemokine binding protein D6, Chemokine receptor CCR 10, Chemokine receptor CCR 9, CMKBR9, D6, hD6
Mouse
Caprylic Acid Precipitation
RUO
1238
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali