missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOT11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10776-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a member of the acyl-CoA thioesterase family which catalyse the conversion of activated fatty acids to the corresponding non-esterified fatty acid and coenzyme A. Expression of a mouse homolog in brown adipose tissue is induced by low temperatures and repressed by warm temperatures. Higher levels of expression of the mouse homolog has been found in obesity-resistant mice compared with obesity-prone mice, suggesting a role of acyl-CoA thioesterase 11 in obesity. Alternative splicing results in transcript variants.Specifica
| ACOT11 | |
| Polyclonal | |
| Unconjugated | |
| ACOT11 | |
| ACOT11, acyl CoA thioesterase 11, BFIT, BFIT1, BFIT2, KIAA0707, STARD14, THEA, THEM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100363074, 26027, 329910 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8VHQ9, Q8WXI4 | |
| Acot11 | |
| ACOT11 Fusion Protein Ag1176 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto