Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Lysate Clearance
Optimized for a variety of applications requiring larger per well volume processing.
Marca: Cytiva (formerly Pall Lab) 8040
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Borosilicate Glass w/o Binder, Supor (PES) | |
Non-sterile | |
10/Pk. |
3.0/1.2 μm | |
Polypropylene Housing | |
350 μL |