Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Optimized for a variety of applications requiring larger per well volume processing.
Marca: Cytiva (formerly Pall Lab) 8048
Dettagli aggiuntivi : Peso : 0.81700kg
Descrizione
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
PTFE | |
Non-sterile | |
10/Pk. |
0.45 μm | |
Solvent Filtration | |
350 μL |