missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Mouse anti-Human, Mouse, Rat, Clone: 3C12C7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66358-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ATCH (adrenocorticotropic hormone) is a hormone which plays a major role in stimulating the adrenal cortex. It is formed through cleavage of the polypeptide precursor proopiomelanocortin (POMC), which also results in several other cleavage products including MSH, ACTH, and beta endorphin. ATCH is secreted from the anterior pituitary in response to the corticotropin-releasing hormone from the hypothalamus. It stimulates the secretion of glucocorticoids like cortisol, but has little control over the stimulation of mineralocorticoids like aldosterone, which is another major hormone of the adrenal cortex.Specifica
| ACTH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P01189, P01193, P01194 | |
| Pomc | |
| ACTH Fusion Protein Ag17311 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 3C12C7 | |
| Unconjugated | |
| POMC | |
| ACTH, ACTH Melanotropin alpha, Adrenocorticotropic hormone, Beta LPH Lipotropin gamma, CLIP, CLIP Lipotropin beta, Corticotropin lipotropin, Gamma LPH Melanotropin beta, LPH, MSH, POC, POMC, Pro opiomelanocortin, proopiomelanocortin | |
| Mouse | |
| Protein G | |
| RUO | |
| 18976, 24664, 5443 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto