Learn More
Epredia™ Lab Vision™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody
Detect muscle-specific actin in immunohistochemistry experiments with Epredia™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-742-S0
Dettagli aggiuntivi : Peso : 0.00170kg
Descrizione
Host Species: Mouse
Clone: HHF35
Isotype: IgG1/κ
Species Reactivity: Human, Rabbit and Rat. Others not known.
Epitope: Not determined
Immunogen: SDS extract of human myocardium
Positive Control: Muscle or sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifica
Actin, Muscle Specific Ab-4 | |
Monoclonal | |
Unconjugated | |
SDS extract of human myocardium | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
HHF35 | |
Mouse | |
100 μL | |
Muscle Biology | |
Human, Rabbit, Rat | |
IgG2a κ |