Learn More
Epredia™ Lab Vision™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody
Detect muscle-specific actin in immunohistochemistry experiments with Epredia™ Actin, Muscle Specific Ab-4, Mouse Monoclonal Antibody.
Marca: Epredia™ MS-742-RQ
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Host Species: Mouse
Clone: HHF35
Isotype: IgG1/κ
Species Reactivity: Human, Rabbit and Rat. Others not known.
Epitope: Not determined
Immunogen: SDS extract of human myocardium
Positive Control: Muscle or sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifica
Actin, Muscle Specific Ab-4 | |
Monoclonal | |
Unconjugated | |
SDS extract of human myocardium | |
IVD | |
Primary |
Immunohistochemistry (Paraffin) | |
HHF35 | |
Mouse | |
12 mL | |
Muscle Biology | |
Ready-to-Use |