Learn More
Eppendorf™ Adapter for 15mL Falcon™ F–35-30-17 Rotor
Designed for use with Eppendorf Rotors
Brand: Eppendorf™ 5702706000
101.00 EUR valido fino al 2024-12-31
Usa il codice promo "23292" per avere il tuo prezzo promozionale.
Additional Details : Peso : 0.03000kg
Description
- Adapter
- For 1 conical tube, 15mL
- For Rotor F-35-30-17
- Set of 10 pcs.
Specifications
Conical Adapter | |
F–35-30-17 Rotor | |
10 Pack |
1 Tube | |
1 |