Learn More
Adenovirus Hexon Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
Marca: Novus Biologicals NB600-1386
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Adenovirus Hexon Polyclonal antibody specifically detects Adenovirus Hexon in Adenovirus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
Specifica
| Adenovirus Hexon | |
| Polyclonal | |
| Unconjugated | |
| Phosphate buffered saline with 0.1% Sodium Azide | |
| Goat | |
| IgG purified | |
| RUO | |
| Primary | |
| Adenovirus, Virus | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 4.0 mg/mL | |
| Western Blot 1:100, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Frozen | |
| Hexon protein; Late protein 2; PII | |
| Gradient purified, disrupted native adenovirus type 5 virions. | |
| 0.5 mL | |
| Virology Bacteria and Parasites | |
| Recognizes numerous Adeno proteins including hexon protein, and reacts with Adenovirus types 1, 2, 3, 5, 6, 7a, 8, 31, 40 & 41. Broad reactivity with other Adenovirus types is expected. Goat anti Adenovirus antibody does not cross-react with Parainfluenza types 1-3, Influenza A & B, RSV, hMPV and Rhinovirus type 16. Goat anti Adenovirus antibody does not react with uninfected Hep-2, LLCMK2 and Vero cells. Adenoviruses are 80-110 nm diameter, non-enveloped icosahedral viruses that can cause respiratory illness, infantile gastroenteritis and other mild disorders such as conjunctivitis. Features of Adenovirus have made it a useful tool for gene transfection and expression in mammalian cells. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Titolo del prodotto
For Research Use Only