Learn More
Adenovirus Hexon Antibody - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
277.00€ - 579.00€
Specifica
| Concentrazione | 4.0 mg/mL |
|---|---|
| Diluizione | Western Blot 1:100, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Frozen |
| Applicazioni | Immunohistochemistry (Frozen), ELISA, Immunocytochemistry, Western Blot, Immunofluorescence |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18096624
|
Novus Biologicals
NB600-1386-0.1ML |
0.1 mL |
277.00€
0.10mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18224425
|
Novus Biologicals
NB600-1386 |
0.5 mL |
579.00€
0.50mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Adenovirus Hexon Protein Polyclonal specifically detects Adenovirus Hexon Protein in Adenovirus, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifica
| 4.0 mg/mL | |
| Immunohistochemistry (Frozen), ELISA, Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Goat | |
| Virology Bacteria and Parasites | |
| Phosphate buffered saline with 0.1% Sodium Azide | |
| IgG | |
| IgG purified | |
| Recognizes numerous Adeno proteins including hexon protein, and reacts with Adenovirus types 1, 2, 3, 5, 6, 7a, 8, 31, 40 & 41. Broad reactivity with other Adenovirus types is expected. Goat anti Adenovirus antibody does not cross-react with Parainfluenza types 1-3, Influenza A & B, RSV, hMPV and Rhinovirus type 16. Goat anti Adenovirus antibody does not react with uninfected Hep-2, LLCMK2 and Vero cells. Adenoviruses are 80-110 nm diameter, non-enveloped icosahedral viruses that can cause respiratory illness, infantile gastroenteritis and other mild disorders such as conjunctivitis. Features of Adenovirus have made it a useful tool for gene transfection and expression in mammalian cells. |
| Western Blot 1:100, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus, Adenovirus | |
| Gradient purified, disrupted native adenovirus type 5 virions. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only