missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ADH5 (Human) Recombinant Protein
Human ADH5 full-length ORF ( AAH14665, 1 a.a. - 374 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000128-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
- Sequence: MANEVIKCKAAVAWEAGKPLSIEEIEVAPPKAHEVRIKIIATAVCHTDAYTLSGADPEGCFPVILGHEGAGIVESVGEGVTKLKAGDTVIPLYIPQCGECKFCLNPKTNLCQKIRVTQGKGLMPDGTSRFTCKGKTILHYMGTSTFSEYTVVADISVAKIDPLAPLDKVCLLGCGISTGYGAAVNTAKLEPGSVCAVFGLGGVGLAVIMGCKVAGASRIIGVDINKDKFARAKEFGATECINPQDFSKPIQEVLIEMTDGGVDYSFECIGNVKVMRAALEACHKGWGVSVVVGVAASGGEIATRPFQLVTGRTWKGTAFGGWKSVESVPKLVSEYMSKKIKVDEFVTHNLSFDEINKAFELMHSGKSIRTVVKI
Specifica
AAH14665 | |
alcohol dehydrogenase 5 (class III), chi polypeptide | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADH5 | |
GST |
128 | |
Wheat germ expression system | |
25 μg | |
ADH-3, ADHX, FDH, GSNOR | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |