missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGAP11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA549230
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
AGAP11 Polyclonal antibody specifically detects AGAP11 in Human samples. It is validated for Western Blot.Specifica
AGAP11 | |
Polyclonal | |
Unconjugated | |
AGAP11 | |
AGAP11; AGAP-11; ankyrin repeat and GTPase domain Arf GTPase activating protein 11; ArfGAP with GTPase domain, ankyrin repeat and PH domain 11; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 11; Centaurin-gamma-like protein KIAA1975; KIAA1975 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
119385 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.42 mg/mL | |
PBS with 0.09% sodium azide | |
Q8TF27 | |
AGAP11 | |
KLH conjugated synthetic peptide between 85-111 amino acids from the N-terminal region of human AGAP11. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |