Learn More
AIF Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech CL488-17984-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The AIF antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human AIF. This antibody recognizes human antigen. The AIF antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifica
| AIF | |
| 1000 μg/mL | |
| AIF, AIFM1, PDCD8 | |
| 20 μL | |
| 9131 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |