Learn More
Abeba™ Air-Cushion 32370 Shoes
Air cushion PU-sole with high slip resistance. Sanitized™ treated.
Marca: Abeba™ 32370/39
Dettagli aggiuntivi : Peso : 0.40000kg
Descrizione
- CE EN ISO 20347:2012 O1 SRA
- Microfiber
- White
- Slip resistance PU outsole sole
- Integrated footbed
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Instep area with elastic band
- Decontaminable
Specifica
CE EN ISO 20347:2012 O1 SRA | |
Microfiber | |
Slip-resistant | |
32370 |
White | |
39 | |
Polyurethane |