Learn More
Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers
Easily replace the air filters on your ultra-low temperature freezer with these replacement parts.
Marca: Thermo Scientific™ AF34567
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
Easily replace the air filters on your ultra-low temperature freezer with Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers.
Replacement air filterSpecifica
For use with Forma 88000, HERAfreeze HFU T, Revco UxF and TSU Series of Upright ULT Freezers. | |
Freezer Air Filter | |
Upright | |
HERAfreeze™ HFU-T, Revco™ UxF, Thermo Scientific™ TSU Series |