missing translation for 'onlineSavingsMsg'
Learn More

ALDH1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17390-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897264

  • 325.38€ / 150µL

Spedizione stimata: 06-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.
Specifica

Specifica

ALDH1L1
Polyclonal
Unconjugated
ALDH1L1
10 FTHFDH, ALDH1L1, FTHFD
Rabbit
Antigen Affinity Chromatography
RUO
107747, 10840, 64392
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75891, P28037, Q8R0Y6
Aldh1l1
ALDH1L1 Fusion Protein Ag11285
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali