Learn More
ALP Monoclonal Antibody (CL1679), Invitrogen™
Mouse Monoclonal Antibody
Marca: Thermo Scientific MA524629
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: MDGKDLSAGA VSAVQCIANP IKLARLVMEK TPHCFLTDQG AAQFAAAMGV PEIPGEKLVT ERNKKRLEKE KHEKGAQKTD CQ Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%. Binds to an epitope located within the peptide sequence TPHCFLTDQGAAQFA as determined by overlapping synthetic peptides.
ALP is found in a variety of tumors but only occasionally in normal tissues. Ovarian cancers are highly positive. ALP is further found in brain tumors, some colonic cancers, some lung cancers, some prostate cancers, uterine cancers, some breast cancers and some thyroid cancers. It was not found in serum.Specifica
ALP | |
Monoclonal | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L266 | |
Asrgl1 | |
Recombinant protein corresponding to Human ALP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
CL1679 | |
Unconjugated | |
Asrgl1 | |
2410004D18Rik; ALP; ALP1; asparaginase and isoaspartyl peptidase 1; asparaginase like 1; asparaginase-like 1; asparaginase-like 1 protein; asparaginase-like protein 1; Asparaginase-like sperm autoantigen; ASRGL1; AU040643; AW060375; Beta-aspartyl-peptidase; CRASH; FLJ22316; Glial asparaginase; Gliap; Hiob; Hiob2; isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP; zgc:103568 | |
Mouse | |
Protein A | |
RUO | |
80150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only