missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ 2000 Ts PLUS SOCO Stitched and Taped
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Dimensione della confezione:
pezzi 45
Descrizione
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals
- Filter 100% of particulates down to 0.01 microns in size.
- Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5
- Ultra Low Linting, reduces rinks of crime scene contamination
- Silicone free, critical in spray painting applications
- Tunnelled elasticated hood, wrists, and ankles to minimize the risk of linting and cross contamination
Specifica
Specifica
| Product Type | Controlled Environments ESD Coveralls |
| Size | Small |
| Sesso | Unisex |
| Materiale | Microporous Non-woven Polyethylene Laminate |
| Colore | White |
| Monouso | Reusable |
| Ankle Style | Elastic |
| Stile polso | Elastic, Finger Loops |
| Stile collo | Con cappuccio |
| Tipo di chiusura | Zipper |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto