Learn More
Ansell™ AlphaTec™ 2000 Ts PLUS Stitched and Taped
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Förpackningsstorlek
pezzi 30
Beskrivelse
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals.
- Filters 100% of particulates down to 0.01 microns in size
- Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5
- Ultra Low Linting, reduced risk of contamination in critical areas
- Silicone free, critical in spray painting applications
- Tunnelled elasticated hood, wrists, and ankles to minimize the risk of linting and cross contamination
Tekniske data
Tekniske data
| Product Type | Controlled Environments ESD Coveralls |
| Size | Large |
| Sesso | Unisex |
| Materiale | Microporous Polyethylene Laminate (Non-woven) |
| Colore | White |
| Monouso | Reusable |
| Ankle Style | Elastic |
| Stile polso | Elastic |
| Stile collo | Con cappuccio |
| Tipo di chiusura | Zipper |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion