Learn More
Ansell AlphaTec™ 3000 Apron
Catalog Number p-7228378
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Stile:
Y30W215
Apron with protective multi-layer barrier fabric that is effective against numerous chemicals.
  • Lightweight and durable
Size Small
Materiale Non-woven Multi-layer Barrier Laminate
Colore Yellow
Da utilizzare con (applicazione) Unexpected Leakages Spills or Other Releases, Loading and Unloading Trucks and Vehicles, Loading Trucks & Vehicles, Product Inspection, Cleaning of Plant and Machines
Peso (sistema metrico) 80 gsm
Stile Y30W215
Tipo di giuntura Welded