Learn More
Ansell AlphaTec 3000 AVANT AIRline
Encapsulated chemical protective suits for use with air-fed/supplied systems.
Marca: Ansell YE30T-00755-04-G05
Dettagli aggiuntivi : Peso : 4.36000kg
Descrizione
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Panoramic visor design ensures a good field of vision for the wearer
- A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
- HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
- Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
- Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress.
Specifica
Yellow | |
Large | |
Y30T755D |
Multi-Layer Non-Woven Barrier Laminate Fabric | |
Welded and Taped | |
80 gsm |