missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 3000 Ultrasonically Welded Model 103
Click to view available options
Size:
3X-Large
4X-Large
5X-Large
Dimensione della confezione:
Pezzo
Descrizione
- Reinforced protection: The AlphaTec™ 3000 Ultrasonically Welded - Model 103 protective suit features multi-layered fabric that ensures an effective barrier against numerous inorganic chemicals and biohazards
- Enhanced features: This protective barrier is further reinforced by its double cuffs, dual-zip systems and double zip enclosure, which extends wear life
- Increased visibility: Thanks to its highly visible bright yellow color, this protective outer garment remains particularly noticeable in low-lit environments, enhancing wearer and workplace safety
- Assured antistatic properties: In addition, it complies with EN 1149-5 antistatic standards
Specifica
Specifica
| Product Type | Chemical Protective Clothing |
| Size | 5X-Large |
| Sesso | Unisex |
| Materiale | Multi-layer Barrier Laminate Fabric |
| Colore | Yellow |
| Monouso | Reusable |
| Stile vita | Elastic |
| Ankle Style | Elastic |
| Stile polso | Elastic, Double |
| Stile collo | Collared |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto