Learn More
Ansell™ AlphaTec™ 3000 Ultrasonically Welded and Taped- Model 103
Codice prodotto. p-200137397
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Codice prodotto. 31114760
Marca: Ansell™ YE30T0010302
Lightweight multi-layer fabric, an effective chemical barrier with dual-zip systems and double cuffs
  • The AlphaTec™ 3000 Ultrasonically Welded & Taped- Model 103 protective suit features multi-layered fabric that ensures an effective barrier against numerous inorganic chemicals and biohazards
  • This protective barrier is further reinforced by its double cuffs, dual-zip systems and double zip enclosure, which extends wear life
  • Highly visible bright yellow color, this protective outer garment remains particularly noticeable in low-lit environments, enhancing wearer and workplace safety
  • In addition, it complies with EN 1149-5 antistatic standards
TRUSTED_SUSTAINABILITY
Specifica
Product Type Chemical Protective Clothing
Size Small
Sesso Unisex
Materiale Multi-layer Barrier Laminate Fabric
Colore Yellow
Monouso Single-use
Stile vita Elastic
Ankle Style Elastic
Stile polso Double, Elastic
Stile collo Collared
Hood Type Standard Collar
Tipo di chiusura Double Zipper
Tipo di giuntura Welded, Taped
Sterilità Non-sterile
Resistenza Chemicals, Anti-static, Particle, Bloodborne Pathogens
Certifications/Compliance (Europe) CE CATEGORY III (Type 3-B, Type 4-B, Type 5-B, Type 6-B, EN 1073-2, EN 14126, EN 1149-5)
Senza lattice Yes
N. di tasche None
Da utilizzare con (applicazione) Transferring Liquids and Solids