Learn More
Euro Protection™ Altaite Low-Cut Safety Shoes
Euro Protection™ Altaite Low-Cut Safety Shoes are comfortable, snug, and durable, and feature a composite toe cap for protection.
Marca: Euro Protection™ 9ALTL44
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Padded collar and bellows tongue for snug fit.
- Heel scuff cap
- Shock-absorbing EVA midsole.
- Full grain leather and bright mesh combination upper.
- Breathable 3D mesh lining.
- Removable, textile and foam insole.
Specifica
Brown, Beige | |
Leather | |
Cemented, shock-absorbing EVA midsole, nitrile rubber outsole, non-metallic (composite fabric) puncture-resistant sole. | |
Low | |
603 g per shoe in size 8 |
Breathable, 3D Mesh | |
44 | |
Nitrile, EVA, Composite Fabric | |
Composite | |
EN ISO 20345 : 2011 |