Learn More
Emma Safety Footwear Amazone XD Safety Shoes
On top of being an extremely safe shoe, Amazone also offers its wearer exceptional comfort. The steel toe cap will protect from falling objects, while the easy twist sole provides a 'sticky grip'.
Marca:  Emma Safety Footwear MM33356645
Dettagli aggiuntivi : Peso : 0.40000kg
Codice prodotto. 15915140
  • 131.00€ / 1 paio
Spedizione stimata: 25-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifica
Specifica
Amazone XD
Oiled Nubuck
45