Learn More
AMER3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561042
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: YSASTQDQRC RDRVQDLSWL RVEPTGLGVQ AWASVEDQPL QLSTEAVEQV AHGSQLDSEP RSAPAARWSS QGHHPESLGL TLNSQQEGGV SASAPEC Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 31%.
AMER3 Polyclonal antibody specifically detects AMER3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifica
AMER3 | |
Polyclonal | |
Unconjugated | |
AMER3 | |
AMER3; APC membrane recruitment protein 3; FAM123C; family with sequence similarity 123C; protein FAM123C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
205147 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N944 | |
AMER3 | |
Recombinant protein corresponding to Human AMER3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only