Learn More
Ammonium nitrate, ACS, 95% min, Thermo Scientific™
In fertilizers, making nitrous oxide
Marca: Thermo Scientific Alfa Aesar 012363.A3
| Quantità | 2kg |
|---|
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificatori chimici
| H4N2O3 | |
| MFCD00011425 | |
| ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills | |
| CHEBI:63038 | |
| [NH4+].[N+](=O)([O-])[O-] |
Specifica
| Ammonium nitrate | |
| 6484-52-2 | |
| 95% min. | |
| MFCD00011425 | |
| Highly soluble in water; soluble in alcohol and liquid ammonia. | |
| [NH4+].[N+](=O)([O-])[O-] | |
| 80.043 | |
| CHEBI:63038 | |
| ACS |
| For analysis ACS | |
| 2kg | |
| H4N2O3 | |
| ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills | |
| DVARTQFDIMZBAA-UHFFFAOYSA-O | |
| azanium;nitrate | |
| 22985 | |
| 80.04 | |
| 1.72 g/mL |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.