Learn More
Abeba™ Anatom 2280 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marca: Abeba™ 2280/48
Dettagli aggiuntivi : Peso : 1.88000kg
Descrizione
- CE EN ISO 20345:2011 S3 SRC
- Microfiber
- White
- Steel toe cap
- Shock absorbant PU midsole
- Metalfree penetration resistance
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- Antistatic
- Cold insulating lining
- Adjustable instep area
- velcro fastening
- Reflective material
- suitable for heel and foot lift
- cushioned ankle support
Specifica
CE EN ISO 20345:2011 S3 SRC | |
Microfiber | |
Slip-resistant | |
2280 |
White | |
48 | |
Polyurethane | |
Steel |