Learn More
Abeba™ Anatom 2616 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marca: Abeba™ 2616/48
Dettagli aggiuntivi : Peso : 1.26600kg
Descrizione
- CE EN ISO 20345:2011 S1 SRC
- Microfiber, perforated
- White
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Adjustable instep area
- Double velcro fastening
- Reflective material
- Reflective material
- Suitable for hygienically demanding areas
Specifica
CE EN ISO 20345:2011 S1 SRC | |
Microfiber | |
Slip-resistant | |
2616 |
White | |
48 | |
Polyurethane | |
Steel |