Learn More
Abeba™ Anatom 32120 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marca: Abeba™ 32120/40
93.35 EUR valido fino al 2024-04-30
Usa il codice promo "21673" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.24700kg
Descrizione
- CE EN ISO 20345:2011 SB A, E SRC
- Microfiber
- White
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3582)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Breathable lining with Silver ions
- Instep area adjustable, with elastic band
- velcro fastening
- suitable for heel and foot lift
- Suitable for hygienically demanding areas
Specifica
CE EN ISO 20345:2011 SB A, E SRC | |
Microfiber | |
Slip-resistant | |
32120 |
White | |
40 | |
Polyurethane | |
Steel |