Learn More
Abeba™ Anatom 32247 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marca: Abeba™ 32247/48
Dettagli aggiuntivi : Peso : 0.95000kg
Descrizione
- CE EN ISO 20345:2011 S1P SRC
- Velours with breathable textile inlays
- Dark blue
- Steel toe cap
- Shock absorbant PU midsole
- Metalfree penetration resistance
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Scratch-resistant coating in toe area
Specifica
CE EN ISO 20345:2011 S1P SRC | |
Velour | |
Slip-resistant | |
32247 |
Blue | |
48 | |
Polyurethane | |
Steel |