Learn More
ABEBA™ Anatom Green/Yellow Safety Sandals S1P
Robust, durable sandals made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
Marca: ABEBA™ 2283/42
Dettagli aggiuntivi : Peso : 1.16000kg
Descrizione
- Comply to EN ISO 20345: 2012, S1P, SRC
- Slip resistant PU double density outsole
- Green/yellow water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable ACC wave insole
Specifica
Green, Yellow | |
42 | |
EN ISO 20345: 2012 S1P SRC | |
Sandals | |
S1P SRC |
Cordura | |
Steel | |
Men's | |
Water Resistant |