Learn More
ANKRD22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553010
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: KTKQNEALVR MLLDAGVEVN ATDCYGCTAL HYACEMKNPS LIPLLLEARA DPTIKNKHGE SSLDIARRLK FSQIELMLRK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 88%.
ANKRD22 is a protein coding gene.Specifica
ANKRD22 | |
Polyclonal | |
Unconjugated | |
ANKRD22 | |
5430429D21Rik; Ankrd22; ankyrin repeat domain 22; ankyrin repeat domain-containing protein 22; D19Ertd675e | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
118932 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VYY1 | |
ANKRD22 | |
Recombinant protein corresponding to Human ANKRD22 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only