Learn More
ANKS4B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560402
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: VALLDKAATA QNIMNPKKVT RLKEQAQKNA RRQIKECERL QEKHQNKMAH TYSKEESGTL SSSKGTFSRS SPSNASAP Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 82%.
As part of the intermicrovillar adhesion complex/IMAC plays a role in epithelial brush border differentiation, controlling microvilli organization and length. Plays a role in assembly of the complex (PubMed:26812018). May play a role in cellular response to endoplasmic reticulum stress. [UniProt]Specifica
ANKS4B | |
Polyclonal | |
Unconjugated | |
ANKS4B | |
ANKS4B; Ankyrin repeat and SAM domain-containing protein 4B; ankyrin repeat and sterile alpha motif domain containing 4B; harmonin-interacting ankyrin repeat-containing protein; harmonin-interacting ankyrin-repeat containing protein; HARP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
257629 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N8V4 | |
ANKS4B | |
Recombinant protein corresponding to Human ANKS4B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only