missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agencinox™ Scatole per sterilizzazione colorate in alluminio anodizzato
Use these boxes for a variety of general laboratory applications. Agencinox™ Anodised Aluminum Colored Sterilization Boxes keep instruments safe and secure during sterilization process.
Marca: Agencinox™ 8036B
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Anodized aluminum colored instrument boxes
- Available in a variety of sizes
Specifica
Blu | |
Rectangular | |
215 mm | |
50 mm |
Aluminum | |
Scatola per sterilizzazione colorata | |
100 mm |