Learn More
Ansell AlphaTec 3000 Coverall, Collar
Codice prodotto. p-10095024
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Dimensione della confezione:
pezzi 20
pezzi 25
Codice prodotto. 16855010
Marca: Ansell YE30W0010302
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Chemical protective suits with a durable, comfortable multi-layer chemical barrier, and Type 3/4/5 protection.
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Lightweight and durable
  • Highly visible bright yellow color for improved worker safety 
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifica
Size Small
Materiale Multi-layer Barrier Laminate Fabric
Colore Yellow
Tipo di giuntura Welded
Certifications/Compliance (Europe) EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003
Modello Alphatec Model 103
Da utilizzare con (applicazione) Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Unexpected Leakages Spills or Other Releases, Opening and Draining Pumps Valves or Lines, Loading and Unloading Trucks and Vehicles, Filling, Blending and Charging of Raw Materials, Product Inspection, Mixing or Loading Chemicals, Cleaning of Plant and Machines
Peso (sistema metrico) 80 gsm
Stile Y30W103