Learn More
Ansell Guanti ultra-igienici Nitrilite™ Silky
Catalog Number 11732343
Clean nitrile glove with very low levels of ionic content and particles feature dexterity, comfort and long wear potential. Designed for use in Class 10 (ISO 4) cleanroom environments such as biotechnology and medical device manufacturing.
  • Made of nitrile polymer
  • Compatible with Class 10 (ISO 4)
  • Very low levels of ionic content and particulate
  • 100% powder free with no plasticizers, silicone or pigments
  • Ergonomically designed
  • Soft and flexible with textured fingers for better feel and grip
  • Rolled beaded cuff for easy donning
  • Ambidextrous