Learn More
Bax Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610982
Descrizione
Proper development and maintenance of homeostasis within tissues is achieved through cell proliferation, differentiation, and apoptosis. Members of the Bcl-2 protein family function to either inhibit (Bcl-2 and Bcl-XL) or promote (Bax and Bak) apoptosis.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifica
| Bax | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human Bax-α aa. 55-178 | |
| 50 μg | |
| Cell Biology | |
| Canine, Human | |
| IgG1 |
| Western Blot | |
| 3 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only.