Learn More
CD107a Mouse anti-Human, APC, Clone: H4A3, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 641581
Descrizione
Reacts with the heavily glycosylated 110kDa lysosomal-associated membrane protein, LAMP-1. LAMP-1 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-1 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 and LAMP-2 (CD107b) are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini.
Host Species: Mouse
Clone: H4A3
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifica
| CD107a | |
| Monoclonal | |
| APC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| H4A3 | |
| LAMP-1 | |
| Human Adult Adherent Peripheral Blood Cells | |
| 100 Tests | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only.