Learn More
CD123 Mouse anti-Human, PE-Cy7, Clone: 7G3, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 560826
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The 7G3 monoclonal antibody specifically reacts with human CD123, the 70 kDa IL-3 Receptor α (IL-3Rα) chain. CD123 associates with CD131, the 120-140 kDa Common β chain to form the IL-3 Receptor Complex. CD131 is shared with the receptors for interleukins IL-5 and GM-CSF. IL-3Rα is expressed on hematopoietic progenitors and plays an important role in hematopoietic progenitor cell growth and differentiation. It is also expressed by mast cells, macrophages and a CD5+ B cell subset. This antibody has been reported to block the binding of 125I-IL-3 to high and low affinity IL-3 receptors. In functional experiments, this antibody was found to inhibit acute myeloid leukemia cell proliferation, basophil histamine release, endothelial cell-mediated IL-8 secretion, and neutrophil transmigration. This antibody has been reported to be useful for immunoprecipitation, Western blot and immunofluorescent staining for flow cytometry. At the Fifth HLDA Workshop, the human IL-3 receptor was designated CD123.
Specifica
CD123 | |
Monoclonal | |
PE-Cyanine7 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
7G3 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human IL-3Ra-transfected cells | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |