Learn More
CD195 Mouse anti-Human, APC-Cy7, Clone: 2D7/CCR5, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 557755
Descrizione
Reacts with the chemokine receptor, CCR5, a seven transmembrane-spanning G protein-associated molecule. CCR5 belongs to the β chemokine receptor family. It is expressed on a subset of T lymphocytes (CD3+, CD45RO+, CD95+). CCR5 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation. It signals a response to at least three chemokines: RANTES and macrophage inflammatory protein-1 (MIP-1) α and β. Additionally, CCR5 has been found to be a co-receptor for macrophage-tropic HIV-1 on CD4+ cells, a characteristic that is important in viral transmission. Reports indicate that individuals who have partial (heterozygous) or complete (homozygous) deletion of the CCR5 allele, demonstrate resistance to HIV infection. This antibody has been shown to block ligand and gp120 binding. It is also able to neutralize HIV infection. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 2D7/CCR5
Isotype: IgG2a, κ
Species Reactivity: Human
Flow Cytometry, Functional Assay, Immunofluorescence
Specifica
| CD195 | |
| Monoclonal | |
| APC-Cyanine7 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Human CCR5 Transfected Cell Line | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| 2D7/CCR5 | |
| CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only.